Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Weiler® Controlled Flare End Brush, ORS Nasco
Supplier: ORS Nasco
804-10306 804-10310 804-10322 804-10302 804-10314
300044-568EA 205.9 USD InStock
300044-568 300042-786 300044-570 300043-652 300042-788
Weiler® Controlled Flare End Brush, ORS Nasco
Brushes
Specifically designed for increased durability when cleaning right angles, corners and fillet welds.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty