Would you like to log in again?
Weiler® Miniature Stem-Mounted End Brush, ORS Nasco
Supplier: ORS Nasco
804-26106 804-26099 804-26114 804-26113 804-26108 804-26098
300050-070EA 12.22 USD InStock
300050-070 300050-072 300050-068 300049-608 300049-606 300050-066
Weiler® Miniature Stem-Mounted End Brush, ORS Nasco
Brushes
Applications include light deburring, blending surface junctures, electronic miniature instrumentation, tool, and die work, hobby crafts, aerospace, jewelry, computer, dental, optics, and other precision industries.
Use with flexible shaft tools and die grinders.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty