Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Weiler® Polyflex Crimped Wire Hollow-End Brush, ORS Nasco
Supplier: ORS Nasco
804-35350 804-35505
300047-454EA 99.68 USD InStock
300047-454 300046-496
Weiler® Polyflex Crimped Wire Hollow-End Brush, ORS Nasco
Brushes
Elastomer gradually wears, exposing consistent short trim for maximum aggression and long life.
Applications include cleaning and finishing recessed areas or I.D.s, die, mold, and tool cleaning and polishing, rubber and plastic flash removal, rust and paint removal, scale and slag removal, I.D. pipe cleaning, carbon cleaning, weld cleaning, deburring, and spot facing. Use with power drills, drill presses, die grinders, and right-angle air die grinders.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty