Would you like to log in again?
Weiler® Vortec Pro Stem Mounted Knot Wire End Brush, ORS Nasco
Supplier: ORS Nasco
804-36286 804-36288 804-36287 804-36251 804-36289 804-36051 804-36050 804-36250
300085-608CS 193.87 USD InStock
300085-608 300085-484 300085-572 300085-574 300085-486 300085-610 300085-612 300085-614
Weiler® Vortec Pro Stem Mounted Knot Wire End Brush, ORS Nasco
Brushes
High impact action for severe applications in tight areas.
  • Brushes can also be used for rubber and plastic removal
  • Remove large burrs and heavy contamination
Applications include paint removal, deburring, and cleaning and finishing recessed areas.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty