Would you like to log in again?
Cartridges, Filters and Accessories for Moldex® 7000, 7800 and 9000 Series Reusable Respirators, Moldex®
Supplier: Moldex
7200 7940 7640 7500 7300 7140 7920 7600 7960
10065-350BG 20.3 USD InStock
10065-350 10065-344 10065-422 10065-364 10065-420 10065-348 10065-346 10065-358 10065-416
Cartridges, Filters and Accessories for Moldex® 7000, 7800 and 9000 Series Reusable Respirators, Moldex®
Respirators Reusable Respirators
The Moldex® 7000, 7800, and 9000 share a full line of cartridges and filters for the toughest of environments and most common applications.
  • P100 Particulate filter disks allow for a wider field of vision due to a low profile design; they are encased in plastic for durability and rough handling
  • P100 Particulate filter disks fit under welding helmets
  • Use the 7940 P100 Particulate Filter Disk as a stand-alone or piggyback with any Moldex® gas/vapor cartridge using the 7920 piggyback adaptor
  • Easy keyless bayonet mount eliminates mis-threading
  • All product and packaging material is 100% PVC-free, for a greener alternative
The Moldex 7140 and 7640 come pre-assembled, but unlike competitive models, they can be easily disassembled. This feature enables the user to replace either the filter or the cartridge or both as they become used up, which helps to save money and reduce waste. With other brands, users are forced to discard the entire unit. COMPLIANCE JUST GOT A WHOLE LOT EASIER. Moldex® designed the 7000 and 7800 half masks and 9000 full face series to be the most user-friendly respirators on the market. These streamlined respirators feature fewer parts, minimal maintenance, and a wider field of vision. They are all ultra-lightweight, durable, and economical, so maximum comfort with extended use is built-in. The award-winning 7000/7800/9000 series add up to simple, comfortable, and economical respiratory protection that’s just plain easy to wear.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty