Would you like to log in again?
Standard Sample Pan Crimper Press, PerkinElmer
Supplier: PerkinElmer
02190048 02190048
101677-284EA 3458.46 USD InStock
101677-284 76474-986
Standard Sample Pan Crimper Press, PerkinElmer
Crimpers, Cappers and Decappers
Used to crimp covers on standard DSC pans of aluminum, gold and copper.
  • Design incorporates a replaceable crimper head
  • Compatible with DSC
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty