Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pigeon PCC (88-104)

Supplier: AnaSpec
AS-60432-1
103005-928EA 239.69 USD InStock
103005-928
Pigeon PCC (88-104)
Proteins and Peptides
PCC (88-104) is a 17-mer peptide fragment of pigeon cytochrome c that stimulates proliferative T cell responses.
Sequence:KAERADLIAYLKQATAK
MW:1890.2 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty