Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

IL-8 Inhibitor

Supplier: AnaSpec
AS-62401
103007-322EA 138.85 USD InStock
103007-322
IL-8 Inhibitor
Proteins and Peptides
This hexapeptide, acetylated on the amino terminus and amidated on the carboxyl terminus, inhibits the specific binding of 125I-IL-8 to neutrophils. IL-8 is a member of the chemokine alpha subfamily that activates neutrophils and is chemotactic for these cells. IL-8 Inhibitor also suppresses the binding of macrophage inflammatory protein 2 (MIP 2) beta to neutrophils.
Sequence:Ac-RRWWCR-NH2
MW:1003.2 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty