Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclo (-GRGDSP) Peptide

Supplier: AnaSpec
AS-61110-5 AS-61110
103006-350EA 118.38 USD InStock
103006-350 103006-352
Cyclo (-GRGDSP) Peptide
Proteins and Peptides
This cyclic peptide is a potent vasodilator. It is more powerful than the linear GRGDSP in changing the vascular tone of arterioles isolated from rat cremaster muscle.
Sequence:GRGDSP, N to C cyclized
MW:569.6 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty