Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glutamic Acid Decarboxylase (GAD65) (524-543)

Supplier: AnaSpec
AS-62757
103007-512EA 228 USD InStock
103007-512
Glutamic Acid Decarboxylase (GAD65) (524-543)
Proteins and Peptides
This is amino acids 524 to 543 fragment of glutamic acid decarboxylase 65 (GAD65). It is one of the first fragments of this islet antigen to induce proliferative T cell responses in the non-obese diabetic (NOD) mouse model of spontaneous autoimmune diabetes. This peptide is a specific, possibly low affinity, stimulus for the spontaneously arising diabetogenic T cell clone BDC2.5. Immunization with p524–543 increases the susceptibility of the NOD mice to type 1 diabetes induced by the adoptive transfer of BDC2.5 T cells.
Sequence:SRLSKVAPVIKARMMEYGTT
MW:2238.7 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty