Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rana Temporaria Temporin A

Supplier: AnaSpec
AS-64831
103008-474EA 160.77 USD InStock
103008-474
Rana Temporaria Temporin A
Proteins and Peptides
Temporin A is a highly hydrophobic antimicrobial peptide amide derived from the frog Rana temporaria. This antimicrobial peptide exerts its effect by inducing the migration of human monocytes, macrophages, and neutrophils, thus modulating the permeability of the microbial membrane to allow passage of various-sized molecules and exhibiting activity against gram-positive bacteria, particularly antibiotic-resistant gram-positive cocci. Temporin A activity is enhanced when used in combination with other antimicrobial agents.
Sequence:FLPLIGRVLSGIL-NH2
MW:1396.8 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty