Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HCAM-2

Supplier: AnaSpec
AS-62058
103007-164EA 628.46 USD InStock
103007-164
HCAM-2
Proteins and Peptides
A pertussis toxin (PTx) enzyme substrate that is labeled with 6-FAM (6-carboxyfluorescein), Abs/Em=494 nm/521 nm.
Sequence:6-FAM-VFDAVTDVIIKNNLKECGLY
MW:2613 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty