Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

[Asp370]-Tyrosinase (368-376)

Supplier: AnaSpec
AS-61456
103006-502EA 206.08 USD InStock
103006-502
[Asp370]-Tyrosinase (368-376)
Proteins and Peptides
This tumor antigenic peptide presented by HLA-A2 antigen to CTLs, is a tyrosinase fragment. Tyrosinase is a membrane-bound protein involved in the melanin synthesis pathway that is expressed by virtually all primary melanoma lesions and by most of metastatic lesions.
Sequence:YMDGTMSQV
MW:1031.2 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty