Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

[Lys(Me3)4]-Histone H3 (1-21)-GGK,Biotin

Supplier: AnaSpec
AS-64192
103007-964EA 571.46 USD InStock
103007-964
[Lys(Me3)4]-Histone H3 (1-21)-GGK,Biotin
Proteins and Peptides
This peptide is a synthetic peptide corresponding to amino acids 1-21 of human histone H3. It is trimethylated at lysine-4 with a C-terminal Gly-Gly linker followed by a biotinylated Lys. The trimethylation of histone H3 at lysine 4 [H3K4(Me3)] shows cell state and lineage potential by differentiating genes that are expressed, poised for expression, or repressed. H3K4(Me3) also labels imprinting control regions.
Sequence:ART-K(Me3)-QTARKSTGGKAPRKQLA-GGK(Biotin)-NH2
MW:2765.2 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty