Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

[Lys(Me3)9]-Histone H3 (1-21)-GGK,Biotin

Supplier: AnaSpec
AS-64628-1
103008-354EA 599.23 USD InStock
103008-354
[Lys(Me3)9]-Histone H3 (1-21)-GGK,Biotin
Proteins and Peptides
This peptide is Histone H3 amino acid residues 1 to 21. It is trimethylated at lysine 9 with a C-terminal GG linker, followed by a biotinylated lysine. The methylation of histone H3 at lysine is an important epigenetic mark of transcriptional silencing a
Sequence:ARTKQTAR-K(Me3)-STGGKAPRKQLA-GGK(Biotin)-NH2
MW:2764.2 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty