Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

[Lys(Ac)23]-Histone H3 (21-44)-GK,Biotin

Supplier: AnaSpec
AS-65241-1
103009-188EA 599.23 USD InStock
103009-188
[Lys(Ac)23]-Histone H3 (21-44)-GK,Biotin
Proteins and Peptides
This peptide is histone H3 (21-43) acetylated at Lys23 with a C-terminal GG linker followed by a biotinylated Lys. Acetylation of histone H3 occurs at Lys14 or Lys23 without preference. Lysine acetylation in histone H3 is associated with transcriptional activation. Provided at >95% peptide purity, this peptide was dissolved in distilled water at 1 mg/ml and re-lyophilized to powder form.
Sequence:AT-K(Ac)-AARKSAPATGGVKKPHRYRPG-GK(Biotin)
MW:2959.5 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty