Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

[Arg(Me1)3]-Histone H4(1-21)-GGK(Biotin), H4R3(Me1), biotin-labeled, Anaspec Inc.

Supplier: Anaspec Inc
AS-64977-1
103008-636EA 628.46 USD InStock
103008-636
[Arg(Me1)3]-Histone H4(1-21)-GGK(Biotin), H4R3(Me1), biotin-labeled, Anaspec Inc.
Proteins and Peptides
This peptide is histone H4 amino acid residues 1 to 21. It is monomethylated at Arg3, with an acetylated N-terminus and C-terminal GG linker, followed by a biotinylated Lys. Methylation of histone H4 at Arg3 is an important step in transcriptional activation and allows for subsequent acetylation of H4 tails by p300. Provided at >90% peptide purity, this peptide was dissolved in distilled water at 1 mg/ml and re-lyophilized to powder form.'
Sequence:Ac-SG-R(Me1)-GKGGKGLGKGGAKRHRKV-GGK(Biotin)
MW:2616.1 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty