Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PKC (Protein Kinase C) Substrate

Supplier: AnaSpec
AS-27183
102996-882EA 247 USD InStock
102996-882
PKC (Protein Kinase C) Substrate
Proteins and Peptides
The peptide sequence, ERMRPRKRQGSVRRRV (cat# 27183-1), corresponds to pseudosubstrate region of the ε-isotype of protein kinase C (PKCε) with an alanine to serine substitution. PKCε exhibits an apparent specificity for the native peptide (Km = 68 µM).
Sequence:ERMRPRKRQGSVRRRV
MW:2067.5 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty