Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tyrosine Kinase Peptide 3

Supplier: AnaSpec
AS-24546
102996-570EA 239.69 USD InStock
102996-570
Tyrosine Kinase Peptide 3
Proteins and Peptides
The native peptide,RRLIEDAEYAARG (cat# 22581), is derived from pp60src, and considered to be a generic substrate for various protein tyrosine kinases. The fluorescent and biotinylated peptides are used to design a variety of assays for PTKs.
Sequence:RRLIEDAE-pY-AARG
MW:1599.7 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty