Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calpain Inhibitor Peptide

Supplier: AnaSpec
AS-60844
103006-034EA 214.85 USD InStock
103006-034
Calpain Inhibitor Peptide
Proteins and Peptides
Calpains are a family of intracellular Ca2+ dependent cysteine proteases. They respond to Ca2+ signals by cleaving many specific proteins, thereby irreversibly modifying their function(s). They are implicated in a variety of Ca2+ regulated cellular processes as well as various pathological phenomena, such as ischemic injury, muscular dystrophy, diabetes, cataract, atherosclerosis, Alzheimer’s disease, and cancer. Calpains represent potential therapeutic targets for drug discovery.
This 27-residue peptide is derived from subdomain 1B of the repetitive domains of calpain, named peptide B27-WT. It is a potent inhibitor of calpain.
Sequence:DPMSSTYIEELGKREVTIPPKYRELLA
MW:3136.6 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty