Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Peptide Mass Spec Standards

Supplier: AnaSpec
AS-60882
103006-094EA 185.62 USD InStock
103006-094
Peptide Mass Spec Standards
Proteins and Peptides
This kit consists of two calibration mixtures for calibrating mass scale in MALDI-TOF or ESI mass spectrometry. Calibration Mixture 1 ranges from 800 to 1800 daltons, and Calibration Mixture 2 from 1800 to 3800 daltons.
Sequence:
MW:
% peak area by HPLC:95
Storage condition:-20° C

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty