Would you like to log in again?
Phosphopeptide Mass Spec Standards
Supplier: AnaSpec
AS-61145
103006-362EA 214.85 USD InStock
103006-362
Phosphopeptide Mass Spec Standards
Proteins and Peptides
This kit contains 6 individually-packed phosphopeptides at 10 ug each. These phosphopeptides can be used for the characterization of affinity purified phosphorylated peptides in liquid chromatography / mass spectrometry. FQ-pS-EEQQQTEDELQDK MW: 2062.0 (Bovine ß-Casein, monophospho cat# 61146, $120/mg) DLDVPIPGRFDRRV-pS-VAAE MW: 2192.4 (PKA Regulatory Subunit II Substrate, cat# 24516, $295/mg) KRP-pS-QRHGSKY-NH2 MW: 1422.5 (UOM9, Phosphorylated PKC Substrate-3, cat# 20294, $175/mg) SFVLNPTNIGM-pS-KSSQGHVTK MW: 2312.6 (DAM1 [221-241] peptide, cat# 61242, $180/mg) TRDIYETDpYYRK MW: 1702.8 (Kinase Domain of Insulin Receptor, cat# 20274 $195/mg) TRDIpYETDpYpYRK MW: 1862.8 (Kinase Domain of Insulin Receptor, cat# 20272 $240/mg)
% peak area by HPLC:95
Storage condition:-20° C
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty