Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bovine Beta-Casein, Monophosphopeptide

Supplier: AnaSpec
AS-61146
103006-364EA 298.15 USD InStock
103006-364
Bovine Beta-Casein, Monophosphopeptide
Proteins and Peptides
Bovine ß-Casein, monophosphopeptide (phospho-Serine) can be used for characterization of affinity purified phosphorylated peptides in liquid chromatography and for the analysis or detection of phosphorylated peptides in mass spectrometry.
Sequence:FQ-pS-EEQQQTEDELQDK
MW:2062 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty