Would you like to log in again?
Weiler® Polyflex® Crimped Wire Cup Brush, ORS Nasco
Supplier: ORS Nasco
Weiler®
804-35406 804-35186
300043-658EA 94.44 USD InStock
300043-658 300043-104
Weiler® Polyflex® Crimped Wire Cup Brush, ORS Nasco
Brushes
These crimped brushes are designed for use with right-angled grinders to remove rust, paint, oxides, and scale.
Applications include heavy-duty cleaning of metal surfaces, cleaning prior to welding, weld scale and corrosion removal, rust and paint removal, roughening, and deburring.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty