Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Weiler® Black Horsehair Fine Sweep Brushes, ORS Nasco
Supplier: ORS Nasco
Weiler®
804-42002 804-42001
300091-890EA 52.95 USD InStock
300091-890 300042-460
Weiler® Black Horsehair Fine Sweep Brushes, ORS Nasco
Brushes
Use to remove dust and fine dirt on smooth surfaces.
  • Horsehair bristles are very durable and resilient
  • Ideal for tile, wood, linoleum, marble, ceramic tile, and smooth concrete
  • Lacquered hardwood block finish
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty