Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Charcoal activated, tested according to Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals
Warning
Synonyms: Carbon activated

05117-1KG
BJ05117-1KGEA 550.23 USD InStock
BJ05117-1KG
Charcoal activated, tested according to Ph. Eur., Fluka™
Charcoal activated


Formula: C
MW: 12.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
Order Now
Specification Test Results
Autoignition Temperature 842 °F
Identity B complying
Alkali soluble colored substances complying
Sulfated ash Max. 5 %
Loss on drying (120°C) Max. 15 %
Acidity or alkalinity complying
Soluble in acid Max. 3 %
Soluble in ethanol Max. 0.5 %
Copper (Cu) Max. 25.0 ppm
Lead (Pb) Max. 10.0 ppm
Zinc (Zn) Max. 25.0 ppm
Sulfide (S) complying
Absorption capacity complying
Fluorescent substances complying
Total bacterial value Max. 1000 KBE/g
Fungi and yeasts Max. 100 KBE/g

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty