Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonia solution 25% (NH₃ basis) in water, Puriss. p.a., Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Ammonium hydroxide

09861-1L
BJ09861-1LEA 147.38 USD InStock
BJ09861-1L
Ammonia solution 25% (NH₃ basis) in water, Puriss. p.a., Fluka™
Ammonia solution
Packed in coated, shock- and leak-protected glass bottle


Formula: NH₃
MW: 35.05 g/mol
Boiling Pt: 37.7 °C (1.013 hPa)
Melting Pt: -92 °C
Density: 0.90 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
UN: 2672
ADR: 8,III
Order Now
Specification Test Results
Assay (acidimetric) 23.0 - 27.0 %
Non-volatile matter Max. 0.0018 %
Density (D 20/4) 0.890 - 0.915
Silver (Ag) Max. 0.02 ppm
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.05 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 1 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.05 ppm
Chromium (Cr) Max. 0.05 ppm
Copper (Cu) Max. 0.1 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.02 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.05 ppm
Molybdenum (Mo) Max. 0.05 ppm
Sodium (Na) Max. 1 ppm
Nickel (Ni) Max. 0.05 ppm
Lead (Pb) Max. 0.05 ppm
Strontium (Sr) Max. 0.1 ppm
Thallium (Tl) Max. 0.05 ppm
Zinc (Zn) Max. 0.1 ppm
Chloride (Cl) Max. 0.5 ppm
Phosphate (PO₄) Max. 0.5 ppm
Sulfate (SO₄) Max. 2 ppm
Sulfide (S) Max. 0.2 ppm
Carbonate (as CO₂) Max. 10 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty