Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Antimony(III) chloride ≥99.0% (RT), Puriss. p.a. ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Antimony trichloride

10774-500G 10774-100G
BJ10774-500GEA 567.58 USD InStock
BJ10774-500G BJ10774-100G
Antimony(III) chloride ≥99.0% (RT), Puriss. p.a. ACS, Fluka™
Antimony (III) chloride


Formula: SbCl₃
MW: 228.12 g/mol
Boiling Pt: 283 °C (1.013 hPa)
Melting Pt: 73.4 °C
Density: 3.140 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011212
CAS Number: 10025-91-9
EINECS: 233-047-2
UN: 1733
ADR: 8,II
Order Now
Specification Test Results
Assay 99.0 - 101.0 %
Insoluble in Chloroform Max. 0.05 %
Arsenic (As) Max. 200 ppm
Calcium (Ca) Max. 50 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 20 ppm
Zinc (Zn) Max. 5 ppm
Sulfate (SO₄) Max. 0.005 %
Vapor Pressure 1 mmHg ( 49 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty