Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anisole 99%, Reagent Grade

Supplier: Honeywell Research Chemicals
Warning
Synonyms: Methoxybenzene Methyl phenyl ether

123226-1L 123226-2.5L
BJ123226-1LEA 194.33 USD InStock
BJ123226-1L BJ123226-2.5L
Anisole 99%, Reagent Grade
Anisole


Formula: C₇H₈O
MW: 108.14 g/mol
Boiling Pt: 154 °C (1.013 hPa)
Melting Pt: -37 °C
Density: 1.00 g/cm³ (20 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00008354
CAS Number: 100-66-3
EINECS: 202-876-1
UN: 2222
ADR: 3,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/96/93/759693.jpg
Order Now
Specification Test Results
Vapor Density 3.7 (vs air)
Assay (GC) Min. 98.5 %
Identity (IR) complying
Vapor Pressure 10 mmHg ( 42.2 °C)
Autoignition Temperature 887 °F

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty