Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Perchloric acid 67-72%, TraceSELECT™ Ultra for ultratrace analysis, Fluka™

Supplier: Honeywell Research Chemicals
Danger

12415-1L 12415-250ML 12415-500ML
BJ12415-250MLEA 556.96 USD InStock
BJ12415-250ML BJ12415-500ML BJ12415-1L
Perchloric acid 67-72%, TraceSELECT™ Ultra for ultratrace analysis, Fluka™
Perchloric acid
TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions


Formula: HClO₄
MW: 100.46 g/mol
Boiling Pt: 203 °C
Melting Pt: -18 °C
Density: 1.66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011325
CAS Number: 7601-90-3
UN: 1873
ADR: 5.1,I
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/22/16640622.jpg
Order Now
Specification Test Results
Assay 67 - 72 %
Silver (Ag) Max. 0.05 ppb
Aluminium (Al) Max. 5 ppb
Arsenic (As) Max. 0.5 ppb
Barium (Ba) Max. 0.1 ppb
Beryllium (Be) Max. 0.1 ppb
Bismuth (Bi) Max. 0.05 ppb
Calcium (Ca) Max. 5 ppb
Cadmium (Cd) Max. 0.05 ppb
Cerium (Ce) Max. 0.05 ppb
Cobalt (Co) Max. 0.1 ppb
Cesium (Cs) Max. 0.05 ppb
Copper (Cu) Max. 0.5 ppb
Dysprosium (Dy) Max. 0.05 ppb
Erbium (Er) Max. 0.05 ppb
Europium (Eu) Max. 0.05 ppb
Iron (Fe) Max. 1 ppb
Gallium (Ga) Max. 0.05 ppb
Gadolinium (Gd) Max. 0.05 ppb
Holmium (Ho) Max. 0.05 ppb
Indium (In) Max. 0.05 ppb
Potassium (K) Max. 5 ppb
Lanthanum (La) Max. 0.05 ppb
Lithium (Li) Max. 0.05 ppb
Lutetium (Lu) Max. 0.05 ppb
Magnesium (Mg) Max. 0.5 ppb
Manganese (Mn) Max. 0.1 ppb
Molybdenum (Mo) Max. 0.1 ppb
Sodium (Na) Max. 10 ppb
Niobium (nb) Max. 0.05 ppb
Neodymium (nd) Max. 0.05 ppb
Nickel (Ni) Max. 0.1 ppb
Lead (Pb) Max. 0.2 ppb
Palladium (Pd) Max. 0.5 ppb
Praseodymium (Pr) Max. 0.05 ppb
Platinum (Pt) Max. 0.1 ppb
Rubidium (Rb) Max. 0.1 ppb
Rhodium (Rh) Max. 0.1 ppb
Antimony (Sb) Max. 0.1 ppb
Samarium (Sm) Max. 0.05 ppb
Tin (Sn) Max. 0.1 ppb
Strontium (Sr) Max. 0.1 ppb
Terbium (Tb) Max. 0.05 ppb
Thorium (Th) Max. 0.1 ppb
Titanium (Ti) Max. 0.5 ppb
Thallium (Tl) Max. 0.05 ppb
Thulium (Tm) Max. 0.05 ppb
Uran (U) Max. 0.05 ppb
Vanadium (V) Max. 0.2 ppb
Yttrium (Y) Max. 0.05 ppb
Ytterbium (Yb) Max. 0.05 ppb
Zinc (Zn) Max. 1 ppb
Zirconium (Zr) Max. 2 ppb

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty