Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetonitrile : water : trifluoroacetic acid (50 : 47.5 : 2.5) %, LabReady™, Solvent Blend for mass spectrometry, Riedel-de Haën™

Supplier: Honeywell Research Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Danger

19182-250ML
BJ19182-250MLEA 159 USD InStock
BJ19182-250ML
Acetonitrile : water : trifluoroacetic acid (50 : 47.5 : 2.5) %, LabReady™, Solvent Blend for mass spectrometry, Riedel-de Haën™
Acetonitrile : water : trifluoroacetic acid
LabReady™ Solvent Blends

  • Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
  • Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
  • Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
  • Honeywell’s high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk


Flash Pt: <10 °C
Storage Temperature: Ambient
CAS Number: 75-05-8, 7732-18-5, 76-05-1
UN: 1648
ADR: 3,II
Order Now
Specification Test Results
Assay trifluoroacetic acid 2.0 - 3.0 %
Water (Karl Fischer) 47 - 49 %
Barium (Ba) Max. 5 ppm
Calcium (Ca) Max. 20 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 20 ppm
Potassium (K) Max. 20 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 20 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty