Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Silver nitrate ≥99.0% ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Lunar causticNitric acid silver saltSilver mononitrate

209139-100G 209139-25G 209139-500G
BJ209139-25GEA 231.44 USD InStock
BJ209139-25G BJ209139-100G BJ209139-500G
Silver nitrate ≥99.0% ACS, Fluka™
Silver nitrate


Formula: AgNO₃
MW: 169.87 g/mol
Boiling Pt: Dec.
Melting Pt: 212 °C
Density: 4.350 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 1493
ADR: 5.1,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/45/16640645.jpg
Order Now
Specification Test Results
Assay Min. 99.0 %
Free acid complying
Copper (Cu) Max. 0.0002 %
Iron (Fe) Max. 0.0002 %
Lead (Pb) Max. 0.001 %
Substances not precipitated by HCl (as SO₄) Max. 0.01 %
Chloride (Cl) Max. 0.0005 %
Sulfate (SO₄) Max. 0.002 %
Appearance of the solution complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty