Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium iodate ≥99.5% ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger

215929-100G 215929-5G
BJ215929-100GEA 113.3 USD InStock
BJ215929-100G BJ215929-5G
Potassium iodate ≥99.5% ACS, Fluka™
Potassium iodate


Formula: KlO₃
MW: 214 g/mol
Melting Pt: 560 °C
Density: 3.93 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011406
CAS Number: 7758-05-6
EINECS: 231-831-9
UN: 1479
ADR: 5.1,II
Order Now
Specification Test Results
Assay 99.4 - 100.4 %
pH (5 %, 25 °C) 5.0 - 8.0
Water insoluble matter Max. 0.005 %
Iron (Fe) Max. 0.001 %
Sodium (Na) Max. 0.005 %
Heavy metals (as Pb) Max. 5 ppm
Bromide, chloride (as Cl) Max. 0.01 %
Iodide (I) Max. 0.001 %
Sulfate (SO₄) Max. 0.005 %
Total N Max. 0.005 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty