Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium hydrogen difluoride 95%, Reagent Grade

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Ammonium bifluoride

224820-2.5KG 224820-500G
BJ224820-2.5KGEA 274.33 USD InStock
BJ224820-2.5KG BJ224820-500G
Ammonium hydrogen difluoride 95%, Reagent Grade
Ammonium hydrogen difluoride


Formula: NH₄HF₂
MW: 57.04 g/mol
Melting Pt: 126 °C (1.013 hPa) (subl.)
Density: 1.500 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012018
CAS Number: 1341-49-7
EINECS: 215-676-4
UN: 1727
ADR: 8,II
Order Now
Specification Test Results
Identity (IR) complying
Assay of NH₄HF₂ 94.0 - 106.0 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty