Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium sulfamate ≥98.0% ACS, Fluka™

Supplier: Honeywell Research Chemicals
Warning
Synonyms: Ammonium sulfamidate

228745-500G 228745-1KG
BJ228745-500GEA 209.04 USD InStock
BJ228745-500G BJ228745-1KG
Ammonium sulfamate ≥98.0% ACS, Fluka™
Ammonium sulphamate


Formula: H₆N₂O₃S
MW: 114.13 g/mol
Melting Pt: 131-135 °C
Density: 1.00 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011429
CAS Number: 7773-06-0
EINECS: 231-871-7
Order Now
Specification Test Results
Identity (IR) complying
Assay Min. 98.0 %
Melting point 131.0 - 135.0 °C
Residue on ignition Max. 0.1 %
Water insoluble matter Max. 0.02 %
Heavy metals (as Pb) Max. 5 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty