Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-2-Butanol ≥99%

Supplier: Honeywell Research Chemicals
Warning
Synonyms: (±)-2-Hydroxy butane (±)-Ethyl methyl carbinol (±)-Methyl ethyl carbinol (±)-sec-Butyl alcohol (±)-sec-Butanol

24126-30KG
BJ24126-30KGEA 733.66 USD InStock
BJ24126-30KG
(±)-2-Butanol ≥99%
(±)-2-Butanol


Formula: H₃CCH₂CH(OH)CH₃
MW: 74.12 g/mol
Boiling Pt: 99 °C
Melting Pt: -89 °C
Density: 0.81 g/cm³ (20 °C)
Flash Pt: 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00004569
CAS Number: 78-92-2
UN: 1120
ADR: 3,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/03/13/710313.jpg
Order Now
Specification Test Results
Assay (GC) Min. 99 %
Non-volatile matter Max. 0.005 %
Water (Karl Fischer) Max. 0.3 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty