Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trichloroethylene ≥99.5% ACS, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Trichloroethene 1,1,2-Trichloroethene 1,1,2-Trichloroethylene

251402-2.5L 251402-1L
BJ251402-2.5LEA 250.43 USD InStock
BJ251402-2.5L BJ251402-1L
Trichloroethylene ≥99.5% ACS, Riedel-de Haën™
Trichloroethylene
ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications


Formula: ClCH=CCl₂
MW: 131.39 g/mol
Boiling Pt: 86-88 °C (1.013 hPa)
Melting Pt: -84 °C
Density: 1.463-1.465 g/cm³ (20 °C)
Flash Pt: No flash point was obtained. but the product may release flammable vapour.
Storage Temperature: Ambient
MDL Number: MFCD00000838
CAS Number: 79-01-6
EINECS: 201-167-4
UN: 1710
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/13/81/741381.jpg
Order Now
Specification Test Results
Assay (GC) Min. 99.50 %
Identity (IR) complying
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.02 %
Titrable Base Max. 0.0003 meq/g
Titrable Acid Max. 0.0001 meq/g
Free halogens complying
Heavy metals (as Pb) Max. 1 ppm
APHA Max. 10
Refractive Index n20/D 1.476 (lit.)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty