Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Perchloric acid ~60% ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger

311413-1L 311413-500ML
BJ311413-500MLEA 261.29 USD InStock
BJ311413-500ML BJ311413-1L
Perchloric acid ~60% ACS, Fluka™
Perchloric acid


Formula: HClO₄
MW: 100.46 g/mol
Boiling Pt: 203 °C (1.013 hPa)
Melting Pt: -18 °C
Density: 1.67 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00011325
CAS Number: 7601-90-3
UN: 1873
ADR: 5.1,I
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/22/16640622.jpg
Order Now
Specification Test Results
Assay 60.0 - 62.0 %
Residue on ignition Max. 0.003 %
Iron (Fe) Max. 1 ppm
Heavy metals (as Pb) Max. 1 ppm
Phosphate, silicate (as SiO₂) Max. 5 ppm
Chloride (Cl) Max. 0.001 %
Sulfate (SO₄) Max. 0.001 %
Total N Max. 0.001 %
APHA Max. 10
Ignition Residue ≤0.003%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty