Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nitric acid concentrate for dilution to 1L, 0.1 M HNO3 (0.1N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Danger

32042-1EA 32042-6X1EA
BJ32042-6X1EAPK 93.93 USD InStock
BJ32042-6X1EA BJ32042-1EA
Nitric acid concentrate for dilution to 1L, 0.1 M HNO3 (0.1N) volumetric solution, Fluka™
Nitric acid
For 1L standard solution


Formula: HNO₃
MW: 63.01 g/mol
Boiling Pt: 100 °C
Melting Pt: no data available
Density: 1.065 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,II
Order Now
Specification Test Results
Factor 0.998 - 1.002
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty