Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl yellow indicator, Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Dimethyl yellowSudan Yellow 4-(Dimethylamino)azobenzene 4-(N,N-Dimethylamino)azobenzene

33129-100G 33129-25G
BJ33129-100GEA 302.09 USD InStock
BJ33129-100G BJ33129-25G
Methyl yellow indicator, Fluka™
Methyl Yellow


Formula: C₁₄H₁₅N₃
MW: 225.29 g/mol
Melting Pt: 111 °C
Storage Temperature: Ambient
MDL Number: MFCD00008308
CAS Number: 60-11-7
EINECS: 200-455-7
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Identity (IR) complying
Change complying
Extinction maximum at 403 - 413 nm
Absorbtion (1%/1cm) 1050 - 1350

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty