Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methanol with 0.1% (v/v) Trifluoroacetic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

34974-4X4L
BJ34974-4X4LCS 780.01 USD InStock
BJ34974-4X4L
Methanol with 0.1% (v/v) Trifluoroacetic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™
Methanol with 0.1% (v/v) Trifluoroacetic acid


Boiling Pt: 64.7 °C (main component)
Melting Pt: -97.68 °C (main component)
Density: 0.792 g/cm³ (20 °C. The physical data is that of the main component)
Flash Pt: 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 76-05-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
Order Now
Specification Test Results
Assay (GC) Min. 99.5 %
Calcium (Ca) Max. 0.5 ppm
Potassium (K) Max. 0.5 ppm
Magnesium (Mg) Max. 0.5 ppm
Sodium (Na) Max. 2 ppm
Transmittance at 210 nm Min. 5 %
Transmittance at 230 nm Min. 35 %
Transmittance from 254 nm Min. 90 %
HPLC Gradient at 254 nm Max. 10 mAU
LC-MS application complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty