Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium hydrogen diiodate ≥99.8% (RT), Puriss. p.a., Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Potassium bi-iodate

60350-100G
BJ60350-100GEA 192.49 USD InStock
BJ60350-100G
Potassium hydrogen diiodate ≥99.8% (RT), Puriss. p.a., Fluka™
Potassium hydrogen diiodate


Formula: HI₂KO₆
MW: 389.91
Storage Temperature: Ambient
MDL Number: MFCD00011400
CAS Number: 13455-24-8
EINECS: 236-650-9
UN: 3085
ADR: 5.1,II
Order Now
Specification Test Results
Assay 99.8 - 100.2 %
Calcium (Ca) Max. 20 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 20 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 20 ppm
Total nitrogen (as N) Max. 20 ppm
Sulfate (SO₄) Max. 50 ppm
Chlorate, Chloride, Bromide, Bromate (as Cl) Max. 200 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty