Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium periodate, dried ≥99.8% (dried basis, RT), Puriss. p.a. ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Potassium metaperiodate Potassium tetroxoiodate (VII)

60450-500G 60450-100G
BJ60450-100GEA 229.08 USD InStock
BJ60450-100G BJ60450-500G
Potassium periodate, dried ≥99.8% (dried basis, RT), Puriss. p.a. ACS, Fluka™
Potassium periodate


Formula: KlO₄
MW: 230 g/mol
Melting Pt: 582 °C
Density: 3.62 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011407
CAS Number: 7790-21-8
EINECS: 232-196-0
UN: 3085
ADR: 5.1,I
Order Now
Specification Test Results
Assay (excluding dried substance) 99.8 - 100.3 %
Loss on drying (110°C) Max. 0.1 %
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 50 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 1 ppm
Sodium (Na) Max. 2000 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Other halogens (as Cl) Max. 100 ppm
Sulfate (SO₄) Max. 50 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty