Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydrogen sulfate monohydrate ≥99.0% (by titrimetric analysis), Puriss. p.a.

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Sodium bisulfate monohydrateSodium bisulphate monohydrate

71657-500G
BJ71657-500GEA 204.27 USD InStock
BJ71657-500G
Sodium hydrogen sulfate monohydrate ≥99.0% (by titrimetric analysis), Puriss. p.a.
Sodium hydrogen sulphate monohydrate


Formula: NaHSO₄·H₂O
MW: 138.08 g/mol
Boiling Pt: Decomposes on heating
Melting Pt: 58 °C
Density: 2.12 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149210
CAS Number: 10034-88-5
Order Now
Specification Test Results
Assay 99.0 - 101.0 %
Silver (Ag) Max. 5 ppm
Aluminium (Al) Max. 5 ppm
Barium (Ba) Max. 5 ppm
Bismuth (Bi) Max. 5 ppm
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 20 ppm
Lithium (Li) Max. 5 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Molybdenum (Mo) Max. 5 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Strontium (Sr) Max. 5 ppm
Thallium (Tl) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Chloride (Cl) Max. 10 ppm
Phosphate (PO₄) Max. 5 ppm
Total N Max. 10 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty