Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Sodium hydrogen selenite ≥96.0% (RT), Purum p.a.

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Sodium biselenite

71658-100G
BJ71658-100GEA 285.09 USD InStock
BJ71658-100G
Sodium hydrogen selenite ≥96.0% (RT), Purum p.a.
Sodium hydrogen selenite
  • Products with a guarantee certificate and/or suitable for the stated analytical application.


Formula: HNaO₃Se
MW: 150.96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003525
CAS Number: 7782-82-3
EINECS: 231-966-3
UN: 2630
ADR: 6.1,I
Order Now
Specification Test Results
Assay 96.0 - 104.0 %
pH (5 %, 20°C) 5.4 - 7.2
Colour of the solution complying
Clearness of the solution complying
Calcium (Ca) Max. 100 ppm
Cadmium (Cd) Max. 50 ppm
Cobalt (Co) Max. 50 ppm
Copper (Cu) Max. 100 ppm
Iron (Fe) Max. 50 ppm
Potassium (K) Max. 100 ppm
Nickel (Ni) Max. 50 ppm
Lead (Pb) Max. 300 ppm
Zinc (Zn) Max. 100 ppm
Chloride (Cl) Max. 100 ppm
Selenate and sulfate (as SO₄) Max. 1500 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty