Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide, tested according to Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Caustic sodaLyeSodium hydrateSoda lye

71694-1KG 71694-5KG
BJ71694-5KGEA 180.22 USD InStock
BJ71694-5KG BJ71694-1KG
Sodium hydroxide, tested according to Ph. Eur., Fluka™
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1.390 °C (1.013 hPa)
Melting Pt: 319 °C
Density: 2.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
Order Now
Specification Test Results
Assay 97.0 - 100.5 %
pH Min. 11.0
Appearance of the solution complying
Iron (Fe) Max. 10 ppm
Heavy metals (as Pb) Max. 20 ppm
Assay of Na₂CO₃ Max. 2 %
Chloride (Cl) Max. 100 ppm
Sulfate (SO₄) Max. 100 ppm
Residual solvents complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty