Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium selenate ≥98.0% (by titrimetric analysis), Purum p.a.

Supplier: Honeywell Research Chemicals
Danger

71948-500G
BJ71948-500GEA 1156.58 USD InStock
BJ71948-500G
Sodium selenate ≥98.0% (by titrimetric analysis), Purum p.a.
Sodium selenate
  • Products with a guarantee certificate and/or suitable for the stated analytical application.


Formula: Na₂O₄Se
MW: 188.94 g/mol
Density: 3.213 g/cm³ (17.5 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003490
CAS Number: 13410-01-0
EINECS: 236-501-8
UN: 2630
ADR: 6.1,I
Order Now
Specification Test Results
Assay 98.0 - 102.0 %
Calcium (Ca) Max. 200 ppm
Cadmium (Cd) Max. 50 ppm
Cobalt (Co) Max. 50 ppm
Copper (Cu) Max. 50 ppm
Iron (Fe) Max. 50 ppm
Potassium (K) Max. 1000 ppm
Nickel (Ni) Max. 50 ppm
Lead (Pb) Max. 50 ppm
Zinc (Zn) Max. 50 ppm
Chloride (Cl) Max. 1000 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty