Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid fuming ≥37% (by titrimetric analysis), TraceSELECT™ for trace analysis, Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Muriatic acid

84415-500ML 84415-100ML
BJ84415-500MLEA 202.64 USD InStock
BJ84415-500ML BJ84415-100ML
Hydrochloric acid fuming ≥37% (by titrimetric analysis), TraceSELECT™ for trace analysis, Fluka™
Hydrochloric acid


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 42 °C (1.013 hPa)
Melting Pt: -35 °C
Density: 1.190 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Assay Min. 37 %
Density (D 20/4) 1.175 - 1.188
Silver (Ag) Max. 0.5 ppb
Aluminium (Al) Max. 5 ppb
Arsenic (As) Max. 5 ppb
Gold (Au) Max. 1 ppb
Barium (Ba) Max. 0.5 ppb
Beryllium (Be) Max. 0.5 ppb
Bismuth (Bi) Max. 0.5 ppb
Calcium (Ca) Max. 10 ppb
Cadmium (Cd) Max. 0.5 ppb
Cobalt (Co) Max. 0.5 ppb
Chromium (Cr) Max. 1 ppb
Cesium (Cs) Max. 0.5 ppb
Copper (Cu) Max. 1 ppb
Iron (Fe) Max. 5 ppb
Gallium (Ga) Max. 0.5 ppb
Indium (In) Max. 0.5 ppb
Iridium (Ir) Max. 0.5 ppb
Potassium (K) Max. 10 ppb
Lithium (Li) Max. 0.5 ppb
Magnesium (Mg) Max. 1 ppb
Manganese (Mn) Max. 0.5 ppb
Molybdenum (Mo) Max. 0.5 ppb
Sodium (Na) Max. 10 ppb
Nickel (Ni) Max. 1 ppb
Lead (Pb) Max. 0.5 ppb
Palladium (Pd) Max. 1 ppb
Platinum (Pt) Max. 0.5 ppb
Rubidium (Rb) Max. 0.5 ppb
Rhodium (Rh) Max. 0.5 ppb
Ruthenium (Ru) Max. 0.5 ppb
Antimony (Sb) Max. 0.5 ppb
Selenium (Se) Max. 1 ppb
Tin (Sn) Max. 1 ppb
Strontium (Sr) Max. 0.5 ppb
Titanium (Ti) Max. 1 ppb
Thallium (Tl) Max. 0.5 ppb
Vanadium (V) Max. 1 ppb
Zinc (Zn) Max. 1 ppb
Zirconium (Zr) Max. 0.5 ppb
Uran (U) Max. 0.5 ppb
Mercury (Hg) Max. 0.5 ppb
Sulfite (SO₃) Max. 1 ppm
Free chlorine (Cl) Max. 0.5 ppm
Bromide (Br) Max. 10 ppm
Phosphate (PO₄) Max. 10 ppb
Sulfate (SO₄) Max. 100 ppb

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty