Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfuric acid solution 49-51% (by titrimetric analysis) for HPLC, Fluka™

Supplier: Honeywell Research Chemicals
Danger

84733-100ML 84733-500ML
BJ84733-100MLEA 145.61 USD InStock
BJ84733-100ML BJ84733-500ML
Sulfuric acid solution 49-51% (by titrimetric analysis) for HPLC, Fluka™
Sulfuric acid


Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: 110 °C (1.013 hPa)
Melting Pt: no data available
Density: 1.290 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 2796
ADR: 8,II
Order Now
Specification Test Results
Assay 49.0 - 51.0 %
Density (D 20/4) 1.375 - 1.405
Absorbance 500 nm Max. 0.02
Absorbance at 240 nm Max. 0.02
Absorbance at 230 nm Max. 0.03
Absorbance at 220 nm Max. 0.04
Absorbance at 210 nm Max. 0.05
Suitability test (HPLC) complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty