Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Toluene ≥99.7% (by GC), Puriss. p.a. ACS, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Benzene, methylMethacideMethane, phenyl-MethylbenzeneMethylbenzolPhenylmethaneToluol

89681-10L 89681-25ML 89681-4X25ML
BJ89681-4X25MLCS 87.93 USD InStock
BJ89681-4X25ML BJ89681-10L BJ89681-25ML
Toluene ≥99.7% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Toluene
ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Beilstein Number: 635760


Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 7.1 %(V)
Melting Pt: -95 °C
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/61/73/766173.jpg
Order Now
Specification Test Results
Vapor Pressure 26mmHg(25°C)
Assay (GC) min. 99.7%
Density (D20/20) 0.865-0.869
Refractive index (n20/D) 1.496-1.498
Non-volatilematter max.0.001%
Water (KarlFischer) max.0.02%
Freeacid (asHCl) max.0.001%
Freealkali (asNaOH) max.0.001%
Aluminium (Al) max.0.00005%
Barium (Ba) max.0.00001%
Bismuth (Bi) max.0.00001%
Calcium (Ca) max.0.00005%
Cadmium (Cd) max.0.000005%
Cobalt (Co) max.0.000002%
Chromium (Cr) max.0.000002%
Copper (Cu) max.0.000002%
Iron (Fe) max.0.00001%
Potassium (K) max.0.00005%
Lithium (Li) max.0.00001%
Magnesium (Mg) max.0.00001%
Manganese (Mn) max.0.000002%
Molybdenum (Mo) max.0.00001%
Sodium (Na) max.0.00005%
Nickel (Ni) max.0.000002%
Lead (Pb) max.0.00001%
Strontium (Sr) max.0.00001%
Tin (Sn) max.0.00001%
Zinc (Zn) max.0.00001%
S-compounds (asS) max.0.003%
Thiophene max.0.0001%
Reaction against H₂SO₄ complying
APHA max.10
VaporDensity 3.2(vsair)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty